Print…
Gilets, Fristads® PR54-521 Design A, royal blue
Fornecedor: FRISTADS KANSAS
8515301312XL 851530131L 851530131M 851530131S 851530131XL 851530131XS 8515302102XL 851530210L 851530210M 851530210S 851530210XL 851530210XS 8515302312XL 851530231L 851530231M 851530231S 851530231XL 851530231XS 8515303312XL 851530331L 851530331M 851530331S 851530331XL 851530331XS 8515304402XL 851530440L 851530440M 851530440S 851530440XL 851530440XS 8515305202XL 851530520L 851530520M 851530520S 851530520XL 851530520XS 8515305302XL 851530530L 851530530M 851530530S 851530530XL 851530530XS 8515305392XL 851530539L 851530539M 851530539S 851530539XL 851530539XS 8515306402XL 851530640L 851530640M 851530640S 851530640XL 851530640XS 8515307302XL 851530730L 851530730M 851530730S 851530730XL 851530730XS 8515309402XL 851530940L 851530940M 851530940S 851530940XL 851530940XS 8515309412XL 851530941L 851530941M 851530941S 851530941XL 851530941XS
113-5286EA 0 EUR
113-5286 113-03759 113-03760 113-03761 113-5285 113-03762 113-5282 113-03751 113-03752 113-03753 113-5281 113-03754 113-5288 113-03763 113-03764 113-03765 113-5287 113-03766 113-5290 113-03767 113-03768 113-03769 113-5289 113-03770 113-5292 113-03771 113-03772 113-03773 113-5291 113-03774 113-5272 113-03731 113-03732 113-03733 113-5271 113-03734 113-5276 113-03739 113-03740 113-03741 113-5275 113-03742 113-5274 113-03735 113-03736 113-03737 113-5273 113-03738 113-5278 113-03743 113-03744 113-03745 113-5277 113-03746 113-5280 113-03747 113-03748 113-03749 113-5279 113-03750 113-5294 113-03775 113-03776 113-03777 113-5293 113-03778 113-5284 113-03755 113-03756 113-03757 113-5283 113-03758
Gilets, Fristads® PR54-521 Design A, royal blue
Uniformes
Gilets made from 65% polyester and 35% cotton; napped inner.
  • Zip fastening
  • 2 breast pockets with flap; front pockets; inner pocket
  • Back pleats; waistband adjustable using press studs, plastic press studs
  • Colour contrasting: Upper collar, bottom edge of zip cover and pocket edges
Material weight: 260 g/m²
Basic colour: Royal blue