Print…
Overalls with bib and braces, Bib'N'Brace 51 FAS
Fornecedor: FRISTADS KANSAS
100310541C146 100310541C148 100310541C150 100310541C152 100310541C154 100310541C156 100310541C44 100310541C46 100310541C48 100310541C50 100310541C52 100310541C54 100310541C56 100310541C58 100310541C60 100310541D100 100310541D104 100310541D108 100310541D112 100310541D116 100310541D120 100310541D96 100310940C146 100310940C148 100310940C150 100310940C152 100310940C154 100310940C156 100310940C44 100310940C46 100310940C48 100310940C50 100310940C52 100310940C54 100310940C56 100310940C58 100310940C60
KWIN100310541C146EA 121.9 EUR
KWIN100310541C146 KWIN100310541C148 KWIN100310541C150 KWIN100310541C152 KWIN100310541C154 KWIN100310541C156 KWIN100310541C44 KWIN100310541C46 KWIN100310541C48 KWIN100310541C50 KWIN100310541C52 KWIN100310541C54 KWIN100310541C56 KWIN100310541C58 KWIN100310541C60 KWIN100310541D100 KWIN100310541D104 KWIN100310541D108 KWIN100310541D112 KWIN100310541D116 KWIN100310541D120 KWIN100310541D96 KWIN100310940C146 KWIN100310940C148 KWIN100310940C150 KWIN100310940C152 KWIN100310940C154 KWIN100310940C156 KWIN100310940C44 KWIN100310940C46 KWIN100310940C48 KWIN100310940C50 KWIN100310940C52 KWIN100310940C54 KWIN100310940C56 KWIN100310940C58 KWIN100310940C60
Overalls with bib and braces, Bib'N'Brace 51 FAS
Uniformes
These bib and brace overalls are made of FAS® twill (100% cotton).
  • Two-way zip down front
  • Cordura® reinforced knee pockets
  • Two tuckable, Cordura® reinforced loose-hanging pockets, one with extra pocket, one with three small pockets and tool loops
  • Two large front and back pockets, folding ruler pocket
  • Leg pocket with tool pocket, en pocket, two loose-hanging chest pockets with fastener
  • Button and loop for sheath knife, two hammer loops, one adjustable
  • OEKO-TEX® certified
Weight: 375 g/m²