Print…
Cotton shirt, long sleeves, Match 210G
Fornecedor: FRISTADS KANSAS
1001179402XL 1001179403XL 100117940L 100117940M 100117940S 100117940XS
113-3594EA 60.9 EUR
113-3594 113-3595 113-3596 113-3597 113-3598 113-3599
Cotton shirt, long sleeves, Match 210G
Uniformes
Shirt for men, manufactured from 100% cotton.
  • Button-down collar
  • Two chest pockets with flaps
  • Snap fasteners
Weight: 210 g/m²