Print…

Tetracycline, Sigma-Aldrich®

Fornecedor: Merck

87128-100G 87128-25G
SIAL87128-100GEA 393 EUR
SIAL87128-100G SIAL87128-25G
Tetracycline, Sigma-Aldrich®
Tetracycline
Fórmula: C₂₂H₂₄N₂O₈
Número MDL : MFCD00151232
Número CAS: 60-54-8
EINECS: 200-481-9

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR