Print…
Dipropyleneglycol monomethyl ether (mixture of isomers), Sigma-Aldrich®
Fornecedor: Merck
Sinônimos: Dipropylene glycol monomethyl ether, 1-((1-Methoxypropan-2-yl)oxy)propan-2-ol
Nível de qualidade M-Clarity™ = MQ200
283282-3L 283282-18L 283282-1L
SIAL283282-3LEA 199 EUR
SIAL283282-3L SIAL283282-18L SIAL283282-1L
Dipropyleneglycol monomethyl ether (mixture of isomers), Sigma-Aldrich®
Dipropyleneglycol monomethyl ether (mixture of isomers)
Fórmula: C₇H₁₆O₃
Peso Molecular: 148.2001 g/mol
Número MDL : MFCD00059604
Número CAS: 34590-94-8
EINECS: 252-104-2