Print…

Ethyl (ethoxymethylene)acetate, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: 3-Ethoxyacrylic acid ethyl ester, Ethyl 3-Ethoxyacrylate, Ethyl 3-Ethoxy-2-Propenoate, Ethyl 3-ethoxyacrylate

Nível de qualidade M-Clarity™ = MQ100

250120-50G 250120-5G
SIAL250120-50GEA 382 EUR
SIAL250120-50G SIAL250120-5G
Ethyl (ethoxymethylene)acetate, Sigma-Aldrich®
Ethyl (ethoxymethylene)acetate
Fórmula: C₇H₁₂O₃
Número MDL : MFCD00009863
Número CAS: 1001-26-9

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR