Print…
Pontas robóticas para PerkinElmer® Janus (Roborack), Axygen®
Fornecedor: Corning
RRP-20-C-R RRP-20-CBK-R RRP-200-C-R RRP-200-CBK-R RRP-50-C-R RRP-50-CBK-R RRF-25-C-R-S RRP-20-CBK-R-S RRP-200-C-R-S RRP-50-C-R-S RRP-50-CBK-R-S RRF-175-C-R-S RRF-175-CBK-RS RRF-25-CBK-R-S
736-0482EA 929.49 EUR
736-0482 736-0480 736-0479 736-0477 736-0484 736-0483 AXYGRRF-25-C-R-S 736-0481 736-0540 736-0543 736-0542 AXYGRRF-175-C-R-S AXYGRRF-175-CBK-RS AXYGRRF-25-CBK-R-S
Pontas robóticas para PerkinElmer® Janus (Roborack), Axygen®
Pontas de pipetas
Transparent or black PP, with or without filter, sterile or non sterile.
  • Black tips are conductive for liquid-level sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase, DNase and endotoxin-free
Rack insert colour: Blue (20 µl)/green (50 µl)/red (200 µl).