Print…
Stirrer shafts with reduced chucking diameter
Fornecedor: Bohlender
C584-52 C592-52 C592-70 C592-64 C592-58 C584-64 C584-58 C581-18 C580-18 C576-22 C576-20 C580-14
BOHLC584-52EA 547 EUR
BOHLC584-52 BOHLC592-52 BOHLC592-70 BOHLC592-64 BOHLC592-58 BOHLC584-64 BOHLC584-58 BOHLC581-18 BOHLC580-18 BOHLC576-22 BOHLC576-20 BOHLC580-14
Stirrer shafts with reduced chucking diameter
Agitadores magnéticos e suspensos Veios agitadores
Stirrer shafts made of PTFE-jacketed stainless steel and the blade/paddle completely made of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
Moon shaped and U-shaped shafts creates tangential flow with little turbulence, whereas all other shafts create axial flow.