Print…
VWR®, Kits de Esponjas para Amostragem de Superfícies
Fornecedor: VWR Chemicals
710-1020EA 812 EUR
710-1020 300-0229 300-0230 300-0231 300-0232 300-0240
VWR®, Kits de Esponjas para Amostragem de Superfícies
Zaragatoas Zaragatoas para colheita de amostras
Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sponge sterile sampling bag, with a foil-sealed, pre-moistened cellulose sponge, and 10 ml of sterile neutralising buffer.
  • Sterilised using gamma irradiation and leakproof transport bags, for the rapid cooling of samples in refrigerators
  • Puncture-proof safety bags eliminating sharp points that can cause puncture and/or abrasions
  • Larger surface in comparison with normal sampling swabs
This surface sampling sponge kit is designed to collect and transport samples, to detect the presence of microbiological contaminations such as Listeria, Salmonella, E. coli, and other food-borne pathogens on almost any surface. These kits are widely used in the food, medical, public health and cosmetic industries.
Certificações: HACCP, USDA, ACIA and CFIA requirements.
Atenção: Gloves should be worn during all sampling procedures.