Print…
Dimethyl glutaconate (mixture of cis and trans isomers), Sigma-Aldrich®
Fornecedor: Merck
Sinônimos: Dimethyl pent-2-enedioate
Nível de qualidade M-Clarity™ = MQ100
D95808-25G D95808-5G
SIALD95808-25GEA 462 EUR
SIALD95808-25G SIALD95808-5G
Dimethyl glutaconate (mixture of cis and trans isomers), Sigma-Aldrich®
Dimethyl glutaconate (mixture of cis and trans isomers)
Fórmula: C₇H₁₀O₄
Peso Molecular: 158.15 g/mol
Número MDL : MFCD00066240
Número CAS: 5164-76-1