Ethyl oxalyl chloride, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos:
Ethyl oxalyl monochloride, Oxalic Acid Monoethyl Ester Chloride, Ethyl Chlorooxoacetate, Ethyl Chloroglyoxylate, Chloroglyoxylic Acid Ethyl Ester, Ethyl chloroformylformate
Nível de qualidade M-Clarity™ = MQ100
E43101-25G
E43101-100G
E43101-500G
SIALE43101-25GEA
38.4
EUR
SIALE43101-25G
SIALE43101-100G
SIALE43101-500G
Ethyl oxalyl chloride, Sigma-Aldrich®
Ethyl oxalyl chloride
Fórmula:
C₄H₅ClO₃ Peso Molecular: 136.53 g/mol |
Número MDL :
MFCD00000706 Número CAS: 4755-77-5 EINECS: 225-285-0 UN: 2920 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...