Print…

Methyl palmitoleate, Sigma-Aldrich®

Fornecedor: Merck


Nível de qualidade M-Clarity™ = MQ200

P9667-100MG P9667-1G P9667-500MG
SIALP9667-100MGEA 109 EUR
SIALP9667-100MG SIALP9667-1G SIALP9667-500MG
Methyl palmitoleate, Sigma-Aldrich®
Methyl palmitoleate
Fórmula: C₁₇H₃₂O₂
Número MDL : MFCD00042911
Número CAS: 1120-25-8
EINECS: 214-303-2

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR