Dimethyl phosphonate, Sigma-Aldrich®
Fornecedor: Merck
Nível de qualidade M-Clarity™ = MQ200
D178454-25G
D178454-500G
SIALD178454-25GEA
83.9
EUR
SIALD178454-25G
SIALD178454-500G
Dimethyl phosphonate, Sigma-Aldrich®
Dimethyl phosphonate
Fórmula:
C₂H₇O₃P |
Número MDL :
MFCD00044633 Número CAS: 868-85-9 EINECS: 212-783-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...