Print…

Octadecyl methacrylate, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: Methacrylic acid Stearyl ester, Octadecyl Methacrylate, Stearyl Methacrylate, Methacrylic acid Octadecyl ester

Nível de qualidade M-Clarity™ = MQ100

411442-1L 411442-250ML
SIAL411442-1LEA 197 EUR
SIAL411442-1L SIAL411442-250ML
Octadecyl methacrylate, Sigma-Aldrich®
Octadecyl methacrylate
Fórmula: C₂₂H₄₂O₂
Peso Molecular: 338.57 g/mol
Número MDL : MFCD00026683
Número CAS: 32360-05-7
EINECS: 251-013-5

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR