Print…
Crotononitrile (mixture of cis and trans isomers), Sigma-Aldrich®
Fornecedor: Merck
Nível de qualidade M-Clarity™ = MQ100
252522-100ML 252522-500ML 252522-5ML
SIAL252522-100MLEA 337 EUR
SIAL252522-100ML SIAL252522-500ML SIAL252522-5ML
Crotononitrile (mixture of cis and trans isomers), Sigma-Aldrich®
Crotononitrile (mixture of cis and trans isomers)
Fórmula: C₄H₅N
Peso Molecular: 67.09 g/mol
Número CAS: 4786-20-3
EINECS: 225-335-1