Print…

Octaethylene glycol, Sigma-Aldrich®

Fornecedor: Merck

15879-1G-F 15879-5G-F
SIAL15879-1G-FEA 166 EUR
SIAL15879-1G-F SIAL15879-5G-F
Octaethylene glycol, Sigma-Aldrich®
Octaethylene glycol
Fórmula: C₁₆H₃₄O₉
Número MDL : MFCD00698694
Número CAS: 5117-19-1
EINECS: 225-856-4

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR