Print…
Colunas capilares, BPX50
Fornecedor: Trajan Scientific and Medical
054740 054750 054751 054752 054761 054772 054741 054742
558-0341EA 456 EUR
558-0341 558-0342 558-0251 568-0446 568-0448 568-0451 568-0445 SGEA054742
Colunas capilares, BPX50
Cromatografia, Colunas
These columns are suitable for EPA methods 604, 608, 8060, 8081, triazines/herbicides, drug screening, steroids and a variety of pharmaceutical applications.
50% phenyl polysilphenylene-siloxane
Max. temperature: 80 °C to 360/370 °C
Suitable replacement for: OV-17, SP-2250, DB-17, DB-17ms, DB-17ht, Rtx-50, SPB-50, HP-50+, HP-17