Print…
Cerium(IV) trifluoromethanesulphonate hydrate, Sigma-Aldrich®
Fornecedor: Merck
422312-10G 422312-1G
SIAL422312-10GEA 355 EUR
SIAL422312-10G SIAL422312-1G
Cerium(IV) trifluoromethanesulphonate hydrate, Sigma-Aldrich®
Cerium(IV) trifluoromethanesulphonate hydrate
Fórmula: C₄CeF₁₂O₁₂S₄·H₂O
Peso Molecular: 754.41 g/mol
Número CAS: 698999-65-4