Chloro(dimethyl)(phenyl)silane, Sigma-Aldrich®
Fornecedor: Merck
Nível de qualidade M-Clarity™ = MQ100
113379-25G
113379-5G
SIAL113379-25GEA
367
EUR
SIAL113379-25G
SIAL113379-5G
Chloro(dimethyl)(phenyl)silane, Sigma-Aldrich®
Chloro(dimethyl)(phenyl)silane
Fórmula:
C₈H₁₁ClSi |
Número MDL :
MFCD00000499 Número CAS: 768-33-2 EINECS: 212-193-0 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...