Print…
Counting chambers, Thoma
Fornecedor: MARIENFELD
0640710 0640711 0640730 0640731
631-0697EA 42.3 EUR
631-0697 720-1898 720-1899 720-1908
Counting chambers, Thoma
Lâminas e lamelas Câmaras de Contagem
Designed for the determination of the yeast cell number and IVD applications.
Certificações: IVD directive 98/79/EC.