Print…
Tubing connectors, Y-shaped, LaboPlast®
Fornecedor: BURKLE
8712-0006 8706-0006 8712-0008 8706-0010 8706-0012 8706-0008 8706-0004 8712-0004 8712-0010 8712-0012 8712-0014 8706-0007 8706-0014
229-7433EA 15.9 EUR
229-7433 229-0128 229-0133 229-0130 229-0131 229-0129 229-0250 229-0200 229-0201 229-0202 229-0203 BURK8706-0007 BURK8706-0014
Tubing connectors, Y-shaped, LaboPlast®
Conexões e Conectores
PP or PVDF.
  • PP, temperature resistant up to +135 °C max.
  • PVDF, temperature resistant up to +160 °C max., highly resistant to chemicals
  • Sterilisable
Cylindrical Y hose connector to connect hoses. PVDF connectors with food industry approval, autoclavable and UV-resistant.