Print…
Recipientes, com tampa roscada, LaboPlast®
Fornecedor: BURKLE
6214-0250 6214-1000 6214-0600 6214-0125 6214-0012 6214-0060 6214-0090 6214-0025
BURK6214-0250EA 1.8 EUR
BURK6214-0250 BURK6214-1000 BURK6214-0600 BURK6214-0125 BURK6214-0012 223-0084 BURK6214-0090 BURK6214-0025
Recipientes, com tampa roscada, LaboPlast®
Recipientes de amostras Recipientes, uso geral
PP, white.
  • Sterilisable
  • Material impermeable to light
  • Suitable for foodstguffs
Shapely packing container with screw cap. Emptying without residues because there are no crevices.