Print…
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos: (2R)-(+)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid, (R)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoic acid
Nível de qualidade M-Clarity™ = MQ100
155268-1G 155268-250MG 155268-5G
SIAL155268-1GEA 275 EUR
SIAL155268-1G SIAL155268-250MG SIAL155268-5G
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, Sigma-Aldrich®
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Fórmula: C₁₀H₉F₃O₃
Número MDL : MFCD00004184
Número CAS: 20445-31-2
EINECS: 243-829-5