Print…
Amostradores de zonas farmacêuticos, Novartos Multi
Fornecedor: BURKLE
5358-0000 5358-0005 5358-0010 5358-0015 5358-0020 5358-1000 5358-0008 5358-0017 5358-0003 5358-0603 5358-1003 5358-1403 5358-0602 5358-1402
300-0187EA 132 EUR
300-0187 300-0188 300-0189 300-0190 300-0191 300-0126 BURK5358-0008 BURK5358-0017 BURK5358-0003 129-1779 129-1780 129-1781 129-1782 129-1783
Amostradores de zonas farmacêuticos, Novartos Multi
Amostradores Amostradores de produtos sólidos
Novartos is the pharmaceutical zone sampler for sampling in accordance with the FDA guidelines for unit dose sampling. With the Novartos pharmaceutical samplers, multiple individual dose samples (unit dose samples) can be taken from one or more sampling points in a single sampling.
  • Sample inserts fit precisely and can easily be replaced
  • Inserts are available in volumes from 0 to 2,0 ml
  • Insertion depths can easily be read from the laser-engraved depth scale
  • Stainless steel AISI 316L (1.4404) / AISI 316Ti (1.4571)
  • Groove-free and hand-polished
  • Easy cleaning
  • 25 mm Ø
Novartos Multi double sampling:
With Novartos Multi double sampling, a double multi-point sample can be taken from multiple predefined sample zones. Depending on the length, four, seven, or nine sample chambers are each fitted with two sample inserts for the double sampling.
Novartos Multi triple sampling:
With Novartos Multi triple sampling, a triple multi-point sample can be taken from multiple predefined sample zones. Depending on the length, four, seven, or nine sample chambers are each fitted with three sample inserts for the triple sampling.
Informações de encomenda: Please order sample inserts separately.