Ethylboronic acid 95%
Fornecedor: Thermo Fisher Scientific
|
Atenção
|
370860010
370860050
ACRO370860010EA
49.4
EUR
ACRO370860010
ACRO370860050
Ethylboronic acid 95%
Ethylboronic acid
Fórmula:
C₂H₇BO₂ Temperatura de Armazenamento: Frigorífico |
Número MDL :
MFCD01074536 Número CAS: 4433-63-0 |
Especificações dos Resultados do Teste
Appearance | White to light yellow Crystalline powder or flakes or needles |
Infrared spectrum | Conforms |
Titration with NaOH | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...