Print…

Methyl isobutyrate, Sigma-Aldrich®

Fornecedor: Merck


Nível de qualidade M-Clarity™ = MQ300

W269409-1KG-K W269409-9KG-K
SIALW269409-1KG-KEA 107 EUR
SIALW269409-1KG-K SIALW269409-9KG-K
Methyl isobutyrate, Sigma-Aldrich®
Methyl isobutyrate
Fórmula: C₅H₁₀O₂
Número MDL : MFCD00008914
Número CAS: 547-63-7
EINECS: 208-929-5
UN: 1237
ADR: 3,II

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR