Print…
Frascos, de boca estreita, redondos, com tampa roscada, WHEATON®
Fornecedor: DWK Life Sciences
209050 209046 208924 208925 208926 208927 208928 209041 209043 209045 209047 209048 209049 209121 209123 209124 209125 209126 209044 208923 209129 209127 209130 209161 209163 209164 209165 209166 209167 209168 209170 209128 209169 W082142B
215-8009EA 190 EUR
215-8009 WHEA209046 WHEA208924 WHEA208925 WHEA208926 WHEA208927 WHEA208928 WHEA209041 WHEA209043 WHEA209045 WHEA209047 WHEA209048 WHEA209049 WHEA209121 WHEA209123 WHEA209124 WHEA209125 WHEA209126 WHEA209044 WHEA208923 WHEA209129 WHEA209127 WHEA209130 WHEA209161 WHEA209163 WHEA209164 WHEA209165 WHEA209166 WHEA209167 WHEA209168 WHEA209170 WHEA209128 WHEA209169 WHEAW082142B
Frascos, de boca estreita, redondos, com tampa roscada, WHEATON®
Frascos Frascos de Plástico
LDPE, HDPE or PP, translucent.
  • Superior double seal closure for leak resistance
  • No-drip pour lip for safe and clean pouring
  • Polypropylene cap attached