Print…
Industrial scales, NewClassic, MS series
Fornecedor: Mettler - Toledo
11124573 11124576 11124588 11124704 11124671 11124683 11124701 11124713 11124722 11124734 11124736 11124740 11124742
611-2498EA 0 EUR
611-2498 611-2499 611-2501 611-2506 611-2503 611-2504 611-2505 611-2507 611-2508 611-2509 METB11124736 611-2510 METB11124742
Industrial scales, NewClassic, MS series
Balanças
A familiar, attractive design of large pan scales that incorporates new technology to deliver rugged, reliable, effective performance. Applications include formulation, statistics, checkweighing, piece counting.
  • MonoBloc weighing technology for fast, repeatable results
  • High grade chemically resistant die-cast aluminium housing
  • USB connectivity to PC for easy transfer of results (on models except KLIPE)
  • Diagnostics check helps audit the balance adjustment history
  • One-touch function via user-defined SmartKeys
Informações de encomenda: OIML approved versions (denoted /M) are available, please ask Avantor for details.
Fornecimento: Supplied with protective cover.
Atenção: * RS232 only