Print…
Agitadores KPG com rosca
Fornecedor: DWK Life Sciences
247500803 247500906
SCOT247500803EA 79.5 EUR
SCOT247500803 SCOT247500906
Agitadores KPG com rosca
Agitadores magnéticos e suspensos Auxiliares de agitação
DURAN®, clear borosilicate glass 3.3.
  • Interchangeable
  • With standard ground joint and GL screw thread tube
  • Ground and polished surface