Print…

Pentadecafluorocaprylic acid, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: Pentadecafluorooctoic acid, Pentadecafluorooctanoic acid, PFOA, Perfluorocaprylic acid, Perfluorooctanoic acid

Nível de qualidade M-Clarity™ = MQ100

171468-25G 171468-5G
SIAL171468-25GEA 152 EUR
SIAL171468-25G SIAL171468-5G
Pentadecafluorocaprylic acid, Sigma-Aldrich®
Pentadecafluorocaprylic acid
Fórmula: C₈HF₁₅O₂
Número MDL : MFCD00004174
Número CAS: 335-67-1
EINECS: 206-397-9
UN: 3261
ADR: 8,III

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR