Print…
Micro burettes, bang pattern, straight stopcock, Schellbach
Fornecedor: HIRSCHMANN
3300140 3300152 3300160 3300240 3300252 3300260
HIRS3300140EA 164 EUR
HIRS3300140 HIRS3300152 HIRS3300160 HIRS3300240 HIRS3300252 HIRS3300260
Micro burettes, bang pattern, straight stopcock, Schellbach
Buretas
Micro burette, Bang pattern, class AS, DURAN®, blue graduation, certified conformity, with dated batch identification.
  • Straight stopcock with standard ground glass key or PTFE key
  • Schellbach stripes for enhanced legibility
  • Main point ring graduation
Certificações: Calibrated for EX, DIN EN ISO 385.