Print…
Burettes, lateral stopcock, class B, amber glass
Fornecedor: HIRSCHMANN
3220160 3220170 3220175
HIRS3220160EA 88 EUR
HIRS3220160 HIRS3220170 HIRS3220175
Burettes, lateral stopcock, class B, amber glass
Buretas
Burette, class B, DURAN®, brown glass.
  • White graduation marks
  • Lateral stopcock with glass key
  • Brown coloured for light sensitive substances
Certificações: Calibrated for EX, DIN EN ISO 385.