Print…
Burettes, Pellet Pattern with intermediate stopcock, SILBERBRAND
Fornecedor: Brand
22336 22338
BRAA22336EA 406 EUR
BRAA22336 BRAA22338
Burettes, Pellet Pattern with intermediate stopcock, SILBERBRAND
Buretas
Amber borosilicate glass 5.4, class B.
  • Calibrated to deliver 'TD, Ex'
  • Automatic zeroing, intermediate stopcock for recirculating residual liquid
25 ml versions: Error limits according to class B, DIN EN ISO 385
50 ml versions: Class B, DIN EN ISO 385
* Reduced distance between subdivision marks.
Total height includes bottle.
Informações de encomenda: Reservoir bottles must be ordered separately.