DL-α-Tocopherol, Sigma-Aldrich®
Fornecedor: Merck
M-Clarity™ quality level = MQ200
SIAL258024-500GEA
660
EUR
SIAL258024-500G
SIAL258024-100G
SIAL258024-5G
DL-α-Tocopherol, Sigma-Aldrich®
DL-α-Tocopherol (Vitamin E)
Fórmula:
C₂₉H₅₀O₂ Peso Molecular: 430,71 g/mol |
Número MDL :
MFCD00072051 Número CAS: 10191-41-0 EINECS: 233-466-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...