Ethyl nitroacetate ≥97%
Fornecedor: Thermo Fisher Scientific
|
Atenção
|
Sinônimos:
Nitroethansäureethylester, Nitroacetic ethyl ester
A14433.06
A14433.14
A14433.22
A14433.06EA
64.8
EUR
A14433.06
A14433.14
A14433.22
Ethyl nitroacetate ≥97%
Ethyl nitroacetate
Purity: 97%
CAS No.: 626-35-7
Molecular Formula: C4H7NO4
Formula Weight: 133.11
Flash Point: 92°(197°F)
Refractive Index: 1.4240
Odour: Weak
Boiling Point: 93-95°/10mm
Beilstein Reference: 1210027
MDL No.: MFCD00007403
Fórmula:
C₄H₇NO₄ Ponto de ebulição: 93…95 °C (10 mmHg) Densidade: 1,202 Ponto de Inflamação: 92 °C (197 °F) Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD00007403 Número CAS: 626-35-7 EINECS: 210-944-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...