Print…

Cadmium acetate hydrate, Sigma-Aldrich®

Fornecedor: Merck
Atenção


Nível de qualidade M-Clarity™ = MQ100

229490-25G 229490-5G
SIAL229490-25GEA 233 EUR
SIAL229490-25G SIAL229490-5G
Cadmium acetate hydrate, Sigma-Aldrich®
Cadmium acetate hydrate
Fórmula: (H₃CCOO)₂Cd·H₂O
Número CAS: 89759-80-8
EINECS: 208-853-2
UN: 2570
ADR: 6.1,III

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR