Print…
1-Bromooctadecane ≥97%
Fornecedor: Thermo Fisher Scientific
A15588.0E A15588.22 A15588.36
A15588.0EEA 268 EUR
A15588.0E A15588.22 A15588.36
1-Bromooctadecane ≥97%
1-Bromooctadecane
Purity: 97% CAS No.: 112-89-0 Molecular Formula: C18H37Br Formula Weight: 333.39 Flash Point: 110°(230°F) Odour: Odourless Boiling Point: 150-152°/0.1mm Beilstein Reference: 774145 MDL No.: MFCD00000231
Fórmula: BrH₂C(CH₂)₁₆CH₃
Número MDL : MFCD00000231
Número CAS: 112-89-0