Print…
Toalhetes para aplicações gerais, WYPALL* L40
Fornecedor: KIMBERLY CLARK
7462 7461 5701 7471
115-0185EA 34.6 EUR
115-0185 KIMB7461 KIMB5701 115-1549
Toalhetes para aplicações gerais, WYPALL* L40
Toalhetes Toalhetes e toalhas descartáveis Toalhetes de papel para aplicações gerais
WYPALL* L40 wipes easily absorb large spills and leaks and are therefore ideal for routine industrial cleaning and maintenance. They are also soft enough to remove soils and liquids from face and hands.
  • Ideal for benchtop use
  • Made of a cellulose/latex mixture
Colour: White.