Print…
VWR® VisiLight® LED3, Fontes de luz fria sem Ventilador
Fornecedor: VWR Collection
631-1100EA 806 EUR
631-1100 631-1103
VWR® VisiLight® LED3, Fontes de luz fria sem Ventilador
Luzes Fontes de Luz Fria
The passively-cooled VisiLight® LED3 cold light source, with fanless cooling system, can be used for a variety of applications, such as quality assurance, industrial endoscopy, machine vision, and is ideal for cleanroom applications. This LED cold light source is characterised by superior finishing quality. Due to the environmentally friendly LED technology, they are durable and perform efficiently. Despite the passive-cooling system, without fan, it easily surpasses the brightness of a 150 W halogen cold light source.
  • Modern design with rugged, anodised aluminium housing
  • Environmentally friendly LED technology with flicker-free illumination
  • 36 W power consumption, luminous intensity (730 lm*) outperforms 150 W halogen
L×P×A: 174×259×81 mm
Peso: 3,7 kg
*com guia de luz ativo Ø=9 mm e L=1 m
*com guia de luz ativo Ø=9 mm e L=1 m
Informações de encomenda: fornecido com uma fonte de alimentação de vários de vários adaptadores (EU, UK, EUA). Tempo de vida do LED de 50 000 horas. Versões noutras temperaturas de cor ou com CRI elevado estão disponíveis a pedido. Contacte-nos para mais informações.