Print…
Funis para pó, de haste curta e larga, de PP
Fornecedor: Brand
148025_U 148035_U 148040_U 148045 148020_U
BRAA148025EA 121 EUR
BRAA148025 BRAA148035 BRAA148040 BRAA148045 BRAA148020
Funis para pó, de haste curta e larga, de PP
Funis Funis para pó
PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
Used for transferring powders. Also suitable for liquids.