Phenoxazine ≥98%
Fornecedor: Thermo Fisher Scientific
L01969.03
L01969.06
L01969.03EA
64.1
EUR
L01969.03
L01969.06
Phenoxazine ≥98%
Phenoxazine
Purity: 98%
CAS No.: 135-67-1
Molecular Formula: C12H9NO
Formula Weight: 183.21
Melting Point: 154-158°
Beilstein Reference: 143234
Merck Reference: 14,7253
MDL No.: MFCD00005014
Fórmula:
C₁₂H₉NO Ponto de ebulição: 180…185 °C (3 mmHg) Ponto de Fusão: 154…158 °C Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD00005014 Número CAS: 135-67-1 EINECS: 205-210-8 Merck Index: 14,7253 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...