Print…
Network vacuum pump units
Fornecedor: VACUUBRAND
733401 737401
181-1058EA 4100 EUR
181-1058 181-1658
Network vacuum pump units
Bombas Bombas de Diafragma
PC 500 LAN NT and PC 600 LAN NT. Chemistry vacuum pump units that are ready for connection, optimised for automatic, demand-sensitive vacuum generation in local vacuum networks e.g. VACUU•LAN®.
  • Chemistry diaphragm pump with high chemical resistance (parts in contact with media are made from PTFE compounds)
  • Oil-free vacuum up to ~9 mbar (PC 500 LAN) or up to ~2 mbar (PC 600 LAN)
  • High suction capacity, even at low suction pressures
  • Gas ballast standard for working with condensates
  • High solvent recovery rate
  • Easy-to-use vacuum controller VNC 1 with digital display
  • If no vacuum is required, the pump unit turns itself off automatically and turns itself back on as required
  • The switching points are freely selectable