Print…
Cold traps with/without condensation drain
Fornecedor: Lenz Laborglas GmbH & CO.KG
05355046 05355558 05355058 05355070 05355570 05355546
181-9982EA 374 EUR
181-9982 181-9981 LENZ05355058 LENZ05355070 LENZ05355570 LENZ05355546
Cold traps with/without condensation drain
Armadilhas de vácuo
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump. DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes