Print…
Autoloop Pro
Fornecedor: WARTEWIG
6.000.360 6.000.210 8.000.400 6.000.400
451-0117EA 64.15 EUR
451-0117 451-0118 WART8.000.400 WART6.000.400
Autoloop Pro
Aquecimentos Bicos de Bunsen /Queimadores de gás
The inoculation loop carrousel Autoloop Pro, which is suitable for up to four inoculation loops, was developed to comply with current requirements in microbiological, cellbiological and biotechnological laboratories.
  • Stable housing enables comfortable and easy access to inoculation loops
  • Reduce gas consumption
  • Intelligent sensor of the Autoloop PRO monitors safe sterilisation
  • Automatic unit switch off after 1 hour
The Autoloop PRO rotates and controls flaming automatically, No unintentional use of hot inoculation loops: When flaming is completed, the display shows the remaining cooling time and the removal positions of cool inoculation loops. All the suspended inoculation loops can be flamed as needed in one full rotation of the carrousel.
Informações de encomenda: Supplied without Fuego SCS series laboratory gas burner.