Print…
VWR®, Tubos de centrífuga, PTFE
Fornecedor: VWR Collection
525-0187EA 28 EUR
525-0187 525-0188 525-0189 525-0190 525-0191 525-0183 525-0184 525-0185 525-0186
VWR®, Tubos de centrífuga, PTFE
Tubos Tubos de centrífuga
De PTFE, superfície macia, com ou sem tampa de rosca.
  • Inert
  • Withstand forces up to 12500×g
  • Excellent chemical resistance
Suitable for use at high and low temperatures (–200 to +280 °C).