Print…
Holders for round bottom flask, silicone, blue
Fornecedor: Saint Gobain Life Sciences
161040 161042 161044
214-0148EA 41.7 EUR
214-0148 214-0149 214-0150
Holders for round bottom flask, silicone, blue
Suportes Suportes de frascos
Can be used to provide a stable support for round-bottomed flasks and other containers with curved bases. Reversible. Non-slip, soft, stackable and easy to clean. Silicone, blue.
  • Good chemical resistance
  • Good thermal and electrical insulation
  • Temperature resistant from −60 to +200 °C