Print…
VWR®, Rolhas para juntas com esmerilado standard
Fornecedor: VWR Collection
217-0210EA 14.6 EUR
217-0210 217-0211 217-0212 217-0213 217-0214 217-0215
VWR®, Rolhas para juntas com esmerilado standard
Rolhas Rolhas de PE/LDPE/HDPE
Greaseless closure, which prevents ground joints from seizing. With inclined, circumferential sealing lips for better sealing and reduced risk of breakage. Easily detachable. With ergonomically shaped grip.
  • PTFE, solid
  • Excellent chemical resistance
  • Gas, liquid and vacuum-tight
  • Temperature resistant from –200 to +300 °C