Print…
Spiral cleanroom notebooks, TexWrite®
Fornecedor: Texwipe
TX5740 TX5741 TX5742
818-0129EA 286 EUR
818-0129 818-0130 818-0121
Spiral cleanroom notebooks, TexWrite®
Blocos de notas
TexWrite® cleanroom spiral-bound notebook, college ruled. An economical choice for note taking and data recording in critical environments.
  • Printed with low sodium, IPA resistant inks
  • Lie flat and pages rotate freely for ease of use
Format: Large (279×216 mm), medium (216×140 mm), small (127×76 mm).
Embalagem: Bag-Within-A-Bag®.