Print…
DURAN® Specimen Jars
Fornecedor: DWK Life Sciences
215802401 215803903 215804805 215805107 215805801
216-3315EA 67.3 EUR
216-3315 216-3316 216-3317 216-3318 SCOT215805801
DURAN® Specimen Jars
Frascos e caixas de coloração Frascos de amostra
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With ground stopper as foot. Air- and liquid-tight if joint grease is used.