Print…
Scoops
Fornecedor: USBECK Laborgeräte
3720 3721 3722 3724 3725 3726 3727 3723
231-0323EA 4.1 EUR
231-0323 231-0324 231-0325 USBE3724 USBE3725 USBE3726 USBE3727 USBE3723
Scoops
Conchas
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use